Gene Transcripts

Gene Symbol
LEMD1
Entrez Gene ID
93273
Gene Name
LEM domain containing 1
Gene Aliases
CT50, LEMP-1
Chromosome Location
Chr. 1: 205381378 - 205449956 on Build GRCh38
UniGene ID
Hs.655520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001552.4 NP_001001552.3 3 232
NM_001199050.1 NP_001185979.1 3 232
XM_011510160.2 XP_011508462.1 3 236
XM_011510161.2 XP_011508463.1 3 224
XM_011510162.2 XP_011508464.1 3 234
XM_011510164.2 XP_011508466.1 3 256
XM_011510165.2 XP_011508467.1 3 252
GenBank mRNA AB085610.1 - 3 199
AB096677.1 - 3 199
AB096680.1 - 3 199
AB096681.1 - 3 199
AK026628.1 - 3 199
AK302392.1 - 3 234
BC036636.2 - 3 202 5296437
DC399934.1 - 3 234

View Full Product Details