Gene Transcripts

Gene Symbol
GRIN3B
Entrez Gene ID
116444
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Gene Aliases
GluN3B, NR3B
Chromosome Location
Chr. 19: 1000419 - 1009814 on Build GRCh38
UniGene ID
Hs.660378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138690.2 NP_619635.1 6 2437
XM_017026243.1 XP_016881732.1 5 1683
GenBank mRNA AY507106.1 - 6 2437
AY507107.1 - 6 2437

View Full Product Details