Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_058167.2 | NP_477515.2 | 7 | 714 | |
NM_194315.1 | NP_919296.1 | 8 | 762 | ||
NM_194457.1 | NP_919439.1 | 6 | 583 | ||
NM_194458.1 | NP_919440.1 | 7 | 843 | ||
XM_005244718.3 | XP_005244775.1 | 7 | 992 | ||
XM_005244719.4 | XP_005244776.1 | 7 | 880 | ||
XM_006710333.3 | XP_006710396.1 | 7 | 1159 | ||
XM_011540613.2 | XP_011538915.1 | 6 | 587 | ||
XM_011540614.2 | XP_011538916.1 | 7 | 945 | ||
XM_011540616.2 | XP_011538918.1 | 8 | 954 | ||
XM_011540617.2 | XP_011538919.1 | 7 | 721 | ||
XM_017000238.1 | XP_016855727.1 | 8 | 825 | ||
XM_017000239.1 | XP_016855728.1 | 6 | 1043 | ||
GenBank mRNA | AF296658.1 | - | 7 | 667 | |
AK056065.1 | - | 7 | 624 | ||
AK075017.1 | - | 7 | 693 | ||
AK098803.1 | - | 2 | 80 | ||
AK291935.1 | - | 7 | 693 | ||
AL555293.3 | - | 8 | 838 | ||
AW411508.1 | - | 3 | 242 | ||
BC094777.1 | - | 7 | 713 | 30340049 | |
BC104890.1 | - | 6 | 538 | 8143893 | |
BC113430.1 | - | 6 | 538 | 8322482 | |
BC143657.1 | - | 6 | 538 | 9052171 | |
BM765278.1 | - | 6 | 498 | ||
BM794599.1 | - | 5 | 453 | ||
BQ881730.1 | - | 5 | 403 |