Gene Transcripts

Gene Symbol
SEPT12
Entrez Gene ID
124404
Gene Name
septin 12
Gene Aliases
SPGF10
Chromosome Location
Chr. 16: 4777602 - 4791610 on Build GRCh38
UniGene ID
Hs.126780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001154458.2 NP_001147930.1 6, 7 850
NM_144605.4 NP_653206.2 7, 8 988
XM_006720846.2 XP_006720909.1 7, 8 1124
XM_011522379.2 XP_011520681.1 6, 7 637
XM_017022938.1 XP_016878427.1 6, 7 1102
GenBank mRNA AK058139.1 - 7, 8 814
BC024017.1 - 7, 8 788
BC035619.1 - 7, 8 794 5172555
DQ456996.1 - 6, 7 676
DQ517531.1 - 7, 8 865
EF620906.1 - 7, 8 988

View Full Product Details