Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080476.4 | NP_536724.1 | 6 | 718 | |
XM_017027664.1 | XP_016883153.1 | 6 | 536 | ||
XM_017027665.1 | XP_016883154.1 | 6 | 536 | ||
GenBank mRNA | AB086842.1 | - | 6 | 524 | |
AK075507.1 | - | 6 | 543 | ||
AK301900.1 | - | 6 | 518 | ||
AK308627.1 | - | 5 | 465 | ||
AY339061.1 | - | 5 | 458 | ||
AY358816.1 | - | 6 | 548 | ||
AY422169.1 | - | 6 | 718 | ||
BC030512.1 | - | 6 | 529 | 4942673 |