Gene Transcripts

Gene Symbol
OR13C4
Entrez Gene ID
138804
Gene Name
olfactory receptor family 13 subfamily C member 4
Gene Aliases
HSHTPCRX17, HTPCRX17, OR2K1, OR37F, OR9-7
Chromosome Location
Chr. 9: 104526253 - 104527209 on Build GRCh38
UniGene ID
Hs.553601
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001919.1 NP_001001919.1 1 601

View Full Product Details