Gene Transcripts
- Gene Symbol
- LINC00477
- Entrez Gene ID
- 144360
- Gene Name
- long intergenic non-protein coding RNA 477
- Gene Aliases
- C12orf67, FAM191B
- Chromosome Location
- Chr. 12: 24566964 - 24584168 on Build GRCh38
- UniGene ID
- Hs.350668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029451.2 |
|
2, 3 |
1338 |
|
GenBank mRNA |
AK057456.1 |
- |
2, 3 |
1338 |
|
View Full Product Details