Gene Transcripts

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
Chromosome Location
Chr. 16: 66598178 - 66696707 on Build GRCh38
UniGene ID
Hs.643961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178818.2 NP_848933.1 3 622
NM_181521.2 NP_852662.1 3 622
XM_011522882.1 XP_011521184.1 3 530
XM_017022954.1 XP_016878443.1 3 526
GenBank mRNA AF479814.1 - 3 622
AF521889.1 - 3 622
AF521890.1 - 3 535
BC093679.1 - 3 504 7939524

View Full Product Details