Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128210.1 | NP_001121682.1 | 2 | 367 | |
NM_181784.2 | NP_861449.2 | 2 | 720 | ||
XM_005264200.4 | XP_005264257.2 | 2 | 797 | ||
XM_005264201.1 | XP_005264258.1 | 2 | 304 | ||
XM_005264202.4 | XP_005264259.1 | 2 | 797 | ||
GenBank mRNA | AK056479.1 | - | 2 | 284 | |
AK299079.1 | - | 2 | 220 | ||
AK307714.1 | - | 2 | 180 | ||
AY299090.1 | - | 2 | 381 | ||
BC029300.1 | - | 2 | 297 | ||
BC111495.1 | - | 2 | 708 | ||
BC130292.1 | - | 2 | 317 | 40146323 | |
BC136334.1 | - | 2 | 317 | 9020321 | |
DC335282.1 | - | 2 | 380 |