Gene Transcripts

Gene Symbol
LHFPL5
Entrez Gene ID
222662
Gene Name
lipoma HMGIC fusion partner-like 5
Gene Aliases
DFNB67, TMHS, dJ510O8.8
Chromosome Location
Chr. 6: 35805294 - 35824075 on Build GRCh38
UniGene ID
Hs.367947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182548.3 NP_872354.1 2 830
GenBank mRNA AK126841.1 - 2 745
BC028630.2 - 2 830 5262164

View Full Product Details