Gene Transcripts

Gene Symbol
RBM20
Entrez Gene ID
282996
Gene Name
RNA binding motif protein 20
Gene Aliases
Chromosome Location
Chr. 10: 110641934 - 110839471 on Build GRCh38
UniGene ID
Hs.116630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134363.2 NP_001127835.2 11 3333
XM_011539697.2 XP_011537999.1 11 3171
XM_017016103.1 XP_016871592.1 11 4591
XM_017016104.1 XP_016871593.1 11 6807
GenBank mRNA AK302802.1 - 1 150
BX648563.1 - 1 198
EU822950.1 - 11 3333

View Full Product Details