Gene Transcripts

Gene Symbol
HCG22
Entrez Gene ID
285834
Gene Name
HLA complex group 22
Gene Aliases
PBMUCL2
Chromosome Location
Chr. 6: 31053450 - 31059890 on Build GRCh38
UniGene ID
Hs.207528
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003948.2 3 1348
GenBank mRNA AB560771.1 - 3 836
AB600644.1 - 3 1344
AK094433.1 - 2 1160

View Full Product Details