Gene Transcripts
- Gene Symbol
- KRTAP8-1
- Entrez Gene ID
- 337879
- Gene Name
- keratin associated protein 8-1
- Gene Aliases
- KAP8.1
- Chromosome Location
- Chr. 21: 30812697 - 30813252 on Build GRCh38
- UniGene ID
- Hs.407655
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175857.3 |
NP_787053.1
|
1 |
478 |
|
View Full Product Details