Gene Transcripts

Gene Symbol
KRTAP5-4
Entrez Gene ID
387267
Gene Name
keratin associated protein 5-4
Gene Aliases
KRTAP5.4
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.539087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012709.1 NP_001012727.1 1084
GenBank mRNA AB126073.1 - 1084

View Full Product Details