Gene Transcripts

Gene Symbol
CCDC184
Entrez Gene ID
387856
Gene Name
coiled-coil domain containing 184
Gene Aliases
C12orf68
Chromosome Location
Chr. 12: 48183583 - 48185926 on Build GRCh38
UniGene ID
Hs.251699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013635.3 NP_001013657.3 1 1746
GenBank mRNA AK056003.1 - 1 1713
AK056783.1 - 1 1220
BC036801.1 - 1 1697 5728402

View Full Product Details