Gene Transcripts

Gene Symbol
TRIM77
Entrez Gene ID
390231
Gene Name
tripartite motif containing 77
Gene Aliases
TRIM77P
Chromosome Location
Chr. 11: 89710299 - 89717872 on Build GRCh38
UniGene ID
Hs.723112
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146162.1 NP_001139634.1 6 1113
NM_001271942.1 NP_001258871.1 5 882
GenBank mRNA JF968446.1 - 5 531

View Full Product Details