Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039141.2 | NP_001034230.1 | 22 | 7132 | |
NM_007032.5 | NP_008963.3 | 12 | 1726 | ||
GenBank mRNA | AB015343.1 | - | 5 | 529 | |
AF281030.1 | - | 12 | 1710 | ||
AL713698.1 | - | 7 | 828 | ||
BC003618.2 | - | 12 | 1648 | ||
BC004303.1 | - | 7 | 1131 | ||
BC013278.2 | - | 7 | 903 | ||
DQ228003.1 | - | 20 | 6795 | ||
DQ228005.1 | - | 21 | 6997 |