Gene Transcripts
- Gene Symbol
- PRICKLE2
- Entrez Gene ID
- 166336
- Gene Name
- prickle planar cell polarity protein 2
- Gene Aliases
- EPM5
- Chromosome Location
- Chr. 3: 64093850 - 64445476 on Build GRCh38
- UniGene ID
- Hs.148105
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198859.3 |
NP_942559.1
|
2, 3 |
736 |
|
XM_011533432.2 |
XP_011531734.1
|
3, 4 |
530 |
|
XM_011533433.2 |
XP_011531735.1
|
3, 4 |
530 |
|
XM_011533434.2 |
XP_011531736.1
|
3, 4 |
355 |
|
XM_011533435.2 |
XP_011531737.1
|
3, 4 |
355 |
|
XM_011533436.2 |
XP_011531738.1
|
2, 3 |
588 |
|
XM_011533437.2 |
XP_011531739.1
|
2, 3 |
9658 |
|
XM_011533440.2 |
XP_011531742.1
|
3, 4 |
530 |
|
XM_017005798.1 |
XP_016861287.1
|
2, 3 |
460 |
|
GenBank mRNA |
AL833539.1 |
- |
2, 3 |
737 |
|
BC119002.1 |
- |
2, 3 |
717 |
40082599 |
View Full Product Details