Gene Transcripts
- Gene Symbol
- GBA
- Entrez Gene ID
- 2629
- Gene Name
- glucosylceramidase beta
- Gene Aliases
- GBA1, GCB, GLUC
- Chromosome Location
- Chr. 1: 155234448 - 155244862 on Build GRCh38
- UniGene ID
- Hs.282997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000157.3 |
NP_000148.2
|
4, 5 |
612 |
|
NM_001005741.2 |
NP_001005741.1
|
5, 6 |
871 |
|
NM_001005742.2 |
NP_001005742.1
|
5, 6 |
852 |
|
NM_001171811.1 |
NP_001165282.1
|
3, 4 |
688 |
|
GenBank mRNA |
AK291911.1 |
- |
5, 6 |
850 |
|
AK300829.1 |
- |
3, 4 |
525 |
|
AK300876.1 |
- |
5, 6 |
852 |
|
AK302000.1 |
- |
3, 4 |
437 |
|
AK311242.1 |
- |
5, 6 |
460 |
|
AK312502.1 |
- |
5, 6 |
668 |
|
AU131071.1 |
- |
5, 6 |
679 |
|
AU140959.1 |
- |
4, 5 |
596 |
|
BC003356.1 |
- |
4, 5 |
551 |
2899915 |
BX648487.1 |
- |
5, 6 |
680 |
|
D13286.1 |
- |
4, 5 |
551 |
|
K02920.1 |
- |
3, 4 |
431 |
|
KJ690771.1 |
- |
4, 5 |
495 |
|
KJ690772.1 |
- |
4, 5 |
495 |
|
KJ690773.1 |
- |
4, 5 |
495 |
|
M16328.1 |
- |
4, 5 |
881 |
|
M19285.1 |
- |
4, 5 |
568 |
|
View Full Product Details