Gene Transcripts

Gene Symbol
OR2A7
Entrez Gene ID
401427
Gene Name
olfactory receptor family 2 subfamily A member 7
Gene Aliases
HSDJ0798C17, OR2A21
Chromosome Location
Chr. 7: 144258696 - 144259628 on Build GRCh38
UniGene ID
Hs.658609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005328.1 NP_001005328.1 1 665
GenBank mRNA BC136704.1 - 1 759
BC136711.1 - 1 759 9020701

Gene Symbol
OR2A4
Entrez Gene ID
79541
Gene Name
olfactory receptor family 2 subfamily A member 4
Gene Aliases
OR2A10
Chromosome Location
Chr. 6: 131699644 - 131701401 on Build GRCh38
UniGene ID
Hs.486488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030908.2 NP_112170.1 1 665
GenBank mRNA BC120953.2 - 1 665
BC120954.1 - 1 665 40115185
BI020419.1 - 1 33

View Full Product Details