Gene Transcripts
- Gene Symbol
- DNAJB3
- Entrez Gene ID
- 414061
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B3
- Gene Aliases
- HCG3
- Chromosome Location
- Chr. 2: 233742750 - 233744015 on Build GRCh38
- UniGene ID
- Hs.725533
- Species
- Homo sapiens
- Gene Symbol
- LOC100286922
- Entrez Gene ID
- 100286922
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B3 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 2: 233754316 - 233755345 on Build GRCh38
- UniGene ID
- Hs.124112
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037694.1 |
|
1 |
270 |
|
NR_037695.1 |
|
1 |
270 |
|
GenBank mRNA |
BC066346.1 |
- |
1 |
255 |
|
View Full Product Details