Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
11593
Gene Name
aspartylglucosaminidase
Gene Aliases
AW060726
Chromosome Location
Chr. 8: 53511687 - 53523422 on Build GRCm38
UniGene ID
Mm.334535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005847.2 NP_001005847.1 6 776
GenBank mRNA BC048230.1 - 7 1737
BC132227.1 - 6 751 40130504
BC132229.1 - 6 751 40130506
BE656262.1 - 2 175
BQ571087.1 - 6 668
S81393.1 - 6 693

View Full Product Details