Gene Transcripts

Gene Symbol
Arsa
Entrez Gene ID
11883
Gene Name
arylsulfatase A
Gene Aliases
AS-A, ASA, AW212749, As-2, As2, TISP73
Chromosome Location
Chr. 15: 89472476 - 89477424 on Build GRCm38
UniGene ID
Mm.620
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009713.4 NP_033843.2 5, 6 2060
GenBank mRNA AK004540.1 - 4, 5 1279
AK029854.1 - 3, 4 1321
AK132501.1 - 4, 5 1291
AK165921.1 - 4, 5 1245
BC011284.1 - 4, 5 1242 3669438
BC098075.1 - 5, 6 2072 30734943
X73230.1 - 4, 5 1493

View Full Product Details