Gene Transcripts
- Gene Symbol
- Capn1
- Entrez Gene ID
- 12333
- Gene Name
- calpain 1
- Gene Aliases
- Capa-1, Capa1, mu-calpin
- Chromosome Location
- Chr. 19: 5988545 - 6015825 on Build GRCm38
- UniGene ID
- Mm.6221
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110504.1 |
NP_001103974.1
|
18, 19 |
1948 |
|
NM_007600.3 |
NP_031626.1
|
18, 19 |
2000 |
|
GenBank mRNA |
AF021847.1 |
- |
18, 19 |
1879 |
|
AF084459.1 |
- |
17, 18 |
1866 |
|
AK088547.1 |
- |
18, 19 |
1920 |
|
AK149090.1 |
- |
14, 15 |
1348 |
|
AK149706.1 |
- |
18, 19 |
1941 |
|
AK150084.1 |
- |
18, 19 |
1939 |
|
AK160940.1 |
- |
18, 19 |
1940 |
|
AK165258.1 |
- |
18, 19 |
1960 |
|
AK168063.1 |
- |
18, 19 |
1939 |
|
AK171405.1 |
- |
18, 19 |
1903 |
|
BC026138.1 |
- |
18, 19 |
1915 |
4218148 |
BC050276.1 |
- |
18, 19 |
1916 |
4923347 |
View Full Product Details