Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007776.2 | NP_031802.2 | 3 | 385 | |
GenBank mRNA | AK136286.1 | - | 3 | 384 | |
BC057013.1 | - | 3 | 366 | 6824399 | |
BC132116.1 | - | 3 | 384 | 40130393 | |
BC132118.1 | - | 3 | 385 | 40130395 | |
CA452140.1 | - | 2 | 232 | ||
K02583.1 | - | 3 | 354 |