Gene Transcripts

Gene Symbol
Dgcr6
Entrez Gene ID
13353
Gene Name
DiGeorge syndrome critical region gene 6
Gene Aliases
Chromosome Location
Chr. 16: 18052860 - 18071632 on Build GRCm38
UniGene ID
Mm.27155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289813.1 NP_001276742.1 4 509
NM_001289814.1 NP_001276743.1 4 500
NM_010047.4 NP_034177.1 5 697
GenBank mRNA AF021031.1 - 4 401
AK019346.1 - 5 700
AK160342.1 - 5 687
BC048503.1 - 4 352 6770478
BE945982.1 - 4 409

View Full Product Details