Gene Transcripts

Gene Symbol
Egfr
Entrez Gene ID
13649
Gene Name
epidermal growth factor receptor
Gene Aliases
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2
Chromosome Location
Chr. 11: 16752203 - 16913907 on Build GRCm38
UniGene ID
Mm.420648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007912.4 NP_031938.1 12 1671
NM_207655.2 NP_997538.1 12 1671
GenBank mRNA AF124513.1 - 12 1605
AF275367.1 - 12 1614
AK004883.1 - 12 1672
AK004911.1 - 12 1646
AK004944.1 - 12 1665
AK134262.1 - 12 1644
BC023729.1 - 12 1642 5351807
U03425.1 - 12 1441
X59698.1 - 12 1550
X78987.1 - 12 1450

View Full Product Details