Gene Transcripts

Gene Symbol
Fut4
Entrez Gene ID
14345
Gene Name
fucosyltransferase 4
Gene Aliases
AI451562, CD15, FAL, FucT-IV, LeX, SSEA-1, Ssea1
Chromosome Location
Chr. 9: 14748459 - 14752122 on Build GRCm38
UniGene ID
Mm.63450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010242.3 NP_034372.1 1 776
GenBank mRNA AK041752.1 - 1 776
BC137588.1 - 1 767 8860607

View Full Product Details