Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162494.1 | NP_001155966.1 | 4 | 1364 | |
NM_008056.3 | NP_032082.3 | 4 | 1368 | ||
GenBank mRNA | AK088399.1 | - | 4 | 1233 | |
AK138964.1 | - | 1 | 81 | ||
BC026150.1 | - | 4 | 1357 | 3983985 | |
U43319.1 | - | 3 | 1194 |