Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207105.3 | NP_996988.2 | 2 | 243 | |
GenBank mRNA | AK077436.1 | - | 2 | 231 | |
AK170370.1 | - | 2 | 225 | ||
AY303784.1 | - | 1 | 76 | ||
AY303785.1 | - | 1 | 67 | ||
BY205244.1 | - | 2 | 262 | ||
M13539.1 | - | 2 | 158 |