Gene Transcripts

Gene Symbol
Dnajb3
Entrez Gene ID
15504
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 3
Gene Aliases
Hsj3, MSJ-1, Msj1
Chromosome Location
Chr. 1: 88204732 - 88205748 on Build GRCm38
UniGene ID
Mm.439983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008299.3 NP_032325.2 1 658
GenBank mRNA AK005688.1 - 1 616
AK005690.1 - 1 607
BC048490.1 - 1 626 6775233
U95607.1 - 1 660

View Full Product Details