Gene Transcripts

Gene Symbol
Olfr55
Entrez Gene ID
100038859
Gene Name
olfactory receptor 55
Gene Aliases
IF6, MOR267-17, MOR267-20
Chromosome Location
Chr. 17: 33176416 - 33177363 on Build GRCm38
UniGene ID
Mm.246543
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010998.2 NP_035128.2 1 533
GenBank mRNA AF102532.1 - 1 344

View Full Product Details