Gene Transcripts

Gene Symbol
Olfr56
Entrez Gene ID
18356
Gene Name
olfactory receptor 56
Gene Aliases
IF7, MOR276-1
Chromosome Location
Chr. 11: 49050733 - 49135387 on Build GRCm38
UniGene ID
Mm.24769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010999.2 NP_035129.1 6 1261
GenBank mRNA AF102533.1 - 1 333
AF102535.1 - 1 333
AF102537.1 - 1 333
AK053352.1 - 6 1261
BC141219.1 - 5 1092 9055831
BC147437.1 - 5 1206 9056030

View Full Product Details