Gene Transcripts

Gene Symbol
Ptprf
Entrez Gene ID
19268
Gene Name
protein tyrosine phosphatase, receptor type, F
Gene Aliases
AA591035, LAR, LARS, RPTP-LAR
Chromosome Location
Chr. 4: 118208213 - 118291400 on Build GRCm38
UniGene ID
Mm.29855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011213.2 NP_035343.2 6 890
GenBank mRNA AF300943.1 - 4 553
AI385919.1 - 2 271

View Full Product Details