Gene Transcripts

Gene Symbol
Sos1
Entrez Gene ID
20662
Gene Name
son of sevenless homolog 1 (Drosophila)
Gene Aliases
4430401P03Rik, 9630010N06, AI449023
Chromosome Location
Chr. 17: 80393752 - 80480453 on Build GRCm38
UniGene ID
Mm.360004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009231.2 NP_033257.2 4 996
GenBank mRNA AK140310.1 - 4 996

View Full Product Details