Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025287.2 | NP_079563.2 | 4 | 334 | |
GenBank mRNA | AB071989.1 | - | 2 | 143 | |
AK004669.1 | - | 4 | 304 | ||
AK028201.1 | - | 3 | 470 | ||
AK030746.1 | - | 3 | 521 | ||
AK132383.1 | - | 3 | 510 | ||
AK159482.1 | - | 3 | 487 | ||
AK160858.1 | - | 3 | 431 | ||
AK166581.1 | - | 3 | 444 | ||
AK168185.1 | - | 3 | 456 | ||
AK168546.1 | - | 3 | 523 | ||
AY538613.1 | - | 2 | 146 | ||
BC043131.1 | - | 3 | 353 | 6811159 | |
BC045205.1 | - | 4 | 304 | 4988521 | |
BC064809.1 | - | 3 | 458 | ||
BY303667.1 | - | 4 | 340 | ||
BY726358.1 | - | 4 | 335 |