Gene Transcripts

Gene Symbol
Hnf1a
Entrez Gene ID
21405
Gene Name
HNF1 homeobox A
Gene Aliases
AI323641, HNF1, HNF1-alpha, HNF1[a], Hnf-1, Hnf1alpha, LFB1, Tcf-1, Tcf1
Chromosome Location
Chr. 5: 114948980 - 114971062 on Build GRCm38
UniGene ID
Mm.332607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009327.3 NP_033353.2 7 1554
GenBank mRNA AK141225.1 - 7 1360
AK144051.1 - 7 1562
BC080698.1 - 7 1556 30471380
M57966.1 - 1534

View Full Product Details