Gene Transcripts

Gene Symbol
Tnp2
Entrez Gene ID
21959
Gene Name
transition protein 2
Gene Aliases
TP2, Tp-2
Chromosome Location
Chr. 16: 10787935 - 10788655 on Build GRCm38
UniGene ID
Mm.206798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013694.4 NP_038722.3 1 257
GenBank mRNA AK005689.1 - 1 262
BC092529.1 - 1 257 6773112
J03494.1 - 1 232

View Full Product Details