Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_194333.3 | NP_919314.2 | 12 | 1687 | |
GenBank mRNA | BC094893.1 | - | 12 | 1690 | |
BC117790.1 | - | 12 | 1674 | 40092152 | |
BC117791.1 | - | 12 | 1674 | 40092153 | |
U25739.1 | - | 12 | 1699 |