Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012034.3 | NP_036164.2 | 4 | 665 | |
GenBank mRNA | AF175523.1 | - | 4 | 566 | |
AK027982.1 | - | 3 | 491 | ||
BC019974.1 | - | 4 | 589 | 4215589 | |
BC052325.1 | - | 4 | 634 | 6531021 |