Gene Transcripts
- Gene Symbol
- Igfbp7
- Entrez Gene ID
- 29817
- Gene Name
- insulin-like growth factor binding protein 7
- Gene Aliases
- AGM, Fstl2, Mac25
- Chromosome Location
- Chr. 5: 77349240 - 77408045 on Build GRCm38
- UniGene ID
- Mm.233470
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159518.1 |
NP_001152990.1
|
3, 4 |
710 |
|
NM_008048.3 |
NP_032074.3
|
2, 3 |
617 |
|
GenBank mRNA |
AB012886.1 |
- |
2, 3 |
618 |
|
AK002277.1 |
- |
2, 3 |
613 |
|
AK148749.1 |
- |
2, 3 |
614 |
|
AK156315.1 |
- |
2, 3 |
619 |
|
BC047202.1 |
- |
2, 3 |
529 |
|
BC092538.1 |
- |
2, 3 |
613 |
30284911 |
BF471478.1 |
- |
3, 4 |
224 |
|
L75822.2 |
- |
2, 3 |
596 |
|
View Full Product Details