Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133926.2 | NP_598687.1 | 7 | 735 | |
GenBank mRNA | AK089742.1 | - | 7 | 718 | |
AK134936.1 | - | 7 | 755 | ||
AK170188.1 | - | 6 | 801 | ||
AK179926.1 | - | 3 | 277 | ||
AK204670.1 | - | 2 | 199 | ||
AK208371.1 | - | 2 | 217 | ||
BC014825.1 | - | 7 | 735 | 3969343 | |
BC042494.1 | - | 3 | 1994 |