Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175095.4 | NP_780304.2 | 3 | 229 | |
GenBank mRNA | AK047941.1 | - | 3 | 239 | |
AK083784.1 | - | 3 | 237 | ||
AK143821.1 | - | 3 | 230 | ||
BC062098.1 | - | 216 | |||
BC100390.1 | - | 3 | 220 | 6439921 | |
BC119316.1 | - | 3 | 223 | 8734065 | |
BC119318.1 | - | 3 | 223 | 8734067 |