Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019913.5 | NP_064297.1 | 2 | 309 | |
GenBank mRNA | AK002358.1 | - | 2 | 284 | |
AK010917.1 | - | 2 | 310 | ||
AK090199.1 | - | 2 | 290 | ||
AK147164.1 | - | 1 | 1237 | ||
AK149855.1 | - | 2 | 293 | ||
AK160591.1 | - | 2 | 312 | ||
AK167925.1 | - | 2 | 247 | ||
AK168322.1 | - | 2 | 275 | ||
BC068182.1 | - | 2 | 295 | 30633447 | |
U85089.1 | - | 2 | 272 |