Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025570.2 | NP_079846.1 | 1, 2 | 121 | |
GenBank mRNA | AB049645.1 | - | 1, 2 | 94 | |
AK004053.1 | - | 1, 2 | 106 | ||
AK011341.1 | - | 1, 2 | 122 | ||
AK015199.1 | - | 105 | |||
AK168665.1 | - | 1, 2 | 95 | ||
BC039985.1 | - | 1, 2 | 91 | 1244957 |