Gene Transcripts

Gene Symbol
Bfar
Entrez Gene ID
67118
Gene Name
bifunctional apoptosis regulator
Gene Aliases
3010001A07Rik, AI666707, AW107665, Bar, Rnf47
Chromosome Location
Chr. 16: 13671858 - 13703612 on Build GRCm38
UniGene ID
Mm.223689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025976.5 NP_080252.1 3 627
GenBank mRNA AK013874.1 - 3 492
AK041839.1 - 3 536
AK178133.1 - 2 339
BC027221.1 - 3 468 4196466

View Full Product Details