Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004359.2 | NP_001004359.1 | 10 | 2915 | |
NM_001005385.1 | NP_001005385.1 | 10 | 2801 | ||
NM_026081.5 | NP_080357.4 | 11 | 3004 | ||
GenBank mRNA | AB093241.1 | - | 5 | 2121 | |
BC027187.1 | - | 1 | 243 | 4505972 | |
BC058970.1 | - | 1 | 1758 | 6826050 | |
BC070413.1 | - | 11 | 2952 | ||
BC085157.1 | - | 10 | 2703 | 30357108 |