Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026157.2 | NP_080433.1 | 9 | 1714 | |
GenBank mRNA | AK002680.1 | - | 9 | 1685 | |
AK011546.1 | - | 9 | 1715 | ||
AK036760.1 | - | 9 | 1719 | ||
AK076565.1 | - | 9 | 1704 | ||
AK135537.1 | - | 9 | 1682 | ||
AK154747.1 | - | 9 | 1715 | ||
AK166038.1 | - | 9 | 1692 | ||
BC017535.1 | - | 9 | 1707 | ||
BC057643.1 | - | 9 | 1688 | 2616306 |