Gene Transcripts

Gene Symbol
Ccdc70
Entrez Gene ID
67929
Gene Name
coiled-coil domain containing 70
Gene Aliases
1700112P19Rik
Chromosome Location
Chr. 8: 21969741 - 21974054 on Build GRCm38
UniGene ID
Mm.60688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026459.3 NP_080735.1 2 285
GenBank mRNA AK006920.1 - 1 178
AK007190.1 - 2 217
BC049553.1 - 2 285 6773824

View Full Product Details