Gene Transcripts

Gene Symbol
Spire1
Entrez Gene ID
68166
Gene Name
spire homolog 1 (Drosophila)
Gene Aliases
6030430B19Rik, AI415299, AU022898, AW550622, Spir-1
Chromosome Location
Chr. 18: 67488209 - 67610902 on Build GRCm38
UniGene ID
Mm.208723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176832.2 NP_789802.1 7 701
NM_194355.2 NP_919336.1 7 799
GenBank mRNA BC058669.1 - 8 841
BC094375.1 - 8 894 30535423

View Full Product Details